missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platelet GPVI Mouse anti-Human, BV480, Clone: HY101 (also known as HY 101∣ HY-101), BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 746734
This item is not returnable.
View return policy
Description
Platelet GPVI Monoclonal antibody specifically detects Platelet GPVI in Human samples. It is validated for Flow Cytometry.
Specifications
| Platelet GPVI | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| HY101 (also known as HY 101| HY-101) | |
| Brilliant Violet 480 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction