missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platelet GPVI Mouse anti-Human, BUV615, Clone: HY101 (also known as HY 101; HY-101), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 751523
This item is not returnable.
View return policy
Description
Platelet GPVI Monoclonal antibody specifically detects Platelet GPVI in Human samples. It is validated for Flow Cytometry
Specifications
| Platelet GPVI | |
| Monoclonal | |
| Brilliant Ultraviolet 615 | |
| Glycoprotein 6; GP6; Platelet glycoprotein VI; Platelet collagen receptor | |
| Glycoprotein VI Recombinant Protein | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| HY101 (also known as HY 101; HY-101) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction