Learn More
Thermo Scientific™ Plate Frame for 96-well Piko PCR Plates, white
Plate Frame for 96-well Piko PCR Plates, white
Brand: Thermo Scientific™ SFR0961
Code : 87
Additional Details : Weight : 0.30000kg
Description
The Thermo Scientific Plate Frame for 96-well PikoPCR Plates, white, is designed for use with the Thermo Scientific Piko PCR Plates, a family of PCR plates which have the footprint of a microscope slide. We have not only shrunk the size of the plate, but dramatically reduced the thickness of the well walls utilizing our patented ultra-thin wall (UTW) technology.Four plates can be snapped into a single frame producing the equivalent of a standard 384-well microplate.
Features
Applications
Related Products
Piko PCR Plate, 96-well, clear
Piko PCR Plate, 96-well, white
Specifications
50 Frame | |
White | |
Piko Thermal Cycler, PikoReal Real-Time PCR System | |
96 |
Piko | |
Frame | |
Polypropylene | |
96-Well Plates |