missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Plain Plates with 0.75mm Spacers
Accessory for use with Verti-GEL Maxi 2-Gel Vertical Gel Electrophoresis System.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.01000kg
Description
- Accessory for use with Verti-GEL Maxi 2-Gel Vertical Gel Electrophoresis System
Specifications
Verti-Gel Maxi 2-Gel Vertical Gel Electrophoresis System | |
Plain Plates with 0.75 mm Spacers |