Learn More
PLAT Rabbit anti-Human, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against recombinant human PLAT.
Marque: Abnova PAB29546.100uL
Description
This gene encodes tissue-type plasminogen activator, a secreted serine protease which converts the proenzyme plasminogen to plasmin, a fibrinolytic enzyme. Tissue-type plasminogen activator is synthesized as a single chain which is cleaved by plasmin to a two chain disulfide linked protein. This enzyme plays a role in cell migration and tissue remodeling. Increased enzymatic activity causes hyperfibrinolysis, which manifests as excessive bleeding; decreased activity leads to hypofibrinolysis which can result in thrombosis or embolism. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: KYSSEFCSTPACSEGNSDCYFGNGSAYRGTHSLTESGASCLPWNSMILIGKVYTAQNPSAQALGLGKHNYCRNPDGDAKPWCHVLKNRRLTWEYCDVPSCSTCGLRQYSQPQFRIKGGLFSpécification
| PLAT | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant human PLAT. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| PLAT | |
| Recombinant protein corresponding to human PLAT. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| DKFZp686I03148/T-PA/TPA | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5327 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.