missing translation for 'onlineSavingsMsg'
Learn More

Plastin L Rabbit anti-Human, Clone: 3T0M1, Novus Biologicals™

Product Code. 18364745 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364745 20 μg 20µL
18344147 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18364745 Supplier Novus Biologicals Supplier No. NBP31645920UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Plastin L Monoclonal antibody specifically detects Plastin L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plastin L
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3T0M1
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 to 4 μg/mL, Immunohistochemistry-Paraffin 1:200-1:500
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CP64, DKFZp781A23186, FLJ25423, FLJ26114, FLJ39956, LC64PPLS2bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)), LCP-1, LPL, L-plastin, L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P), Lymphocyte cytosolic protein 1, lymphocyte cytosolic protein 1 (L-plastin), Lymphocyte cytosolic protein-1 (plasmin), plastin 2, plastin-2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 528-627 of human Plastin L (P13796).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3936
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.