Learn More
Kartell™ Plastilab™ Untapered Y-Shaped Tubing Connectors
BIOPHARMA
Product Code. 12599656
Click to view available options
:
Y-shaped; Bore Size: 2.7mm; 100 Pack
Product Code. 12599656
Brand: Kartell™ 0046500
Only null left
Add to basket
Add to basket
Designed for use with hose system and vacuum lines.
  • Made from polypropylene
  • Autoclavable
  • Nominal dimensions
Compliance
Meet Food and Agriculture regulations
Specifications
Certifications/Compliance Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) Useful for any hoses system and vacuum line, suitable for food stuff
Packaging Quantity 100/Pk.
Port Size 2.7mm bore
Shape Y
Product Type Untapered Y Tubing Connector