Learn More
Kartell™ Plastilab™ Conical Stoppers
Features tapered conical-shaped bottom to ensure sealing action.
Brand:  Kartell™ 0124200
Code : NEW
Additional Details : Weight : 0.00270kg
 View more versions of this product
Product Code. 11346364
  • £1.24 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Made from Polyethylene for tight and secure seal
  • Raised embossed easy-to-read sizes
  • Color-coded
Specifications
Specifications
Conical bottom
Stopper
Polyethylene
DIN 12242
Green
10.4 mm
NS 12/21
Pentagonal top