Learn More
Kartell™ Plastilab™ Conical Stoppers
Product Code. 11336364
Change view
Click to view available options
Diameter (Metric) Bottom:
8.1 mm
10.4 mm
12.2 mm
16.4 mm
25.8 mm
Color:
Black
Blue
Brown
Green
Yellow
Standard Taper Joint:
NS 10/19
NS 12/21
NS 14/23
NS 19/26 Cone
NS 29/32
Unit Size:
Each
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Bottom Color Standard Taper Joint unitSize
11336364 8.1 mm Blue NS 10/19 Each
11366364 16.4 mm Yellow NS 19/26 Cone Each
11356364 12.2 mm Brown NS 14/23 Each
10326161 25.8 mm Black NS 29/32 Pack of 10
11346364 10.4 mm Green NS 12/21 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11336364 Supplier Kartell™ Supplier No. 0124100
Only null left
Add to basket
Add to basket
Features tapered conical-shaped bottom to ensure sealing action.
  • Made from Polyethylene for tight and secure seal
  • Raised embossed easy-to-read sizes
  • Color-coded
Specifications
Certifications/Compliance DIN 12242
Description Conical bottom
Standard Taper Joint NS 10/19
Color Blue
Diameter (Metric) Bottom 8.1 mm
Material Polyethylene
Product Type Tapered Stopper
Shape Pentagonal Grip