Learn More
Kartell™ Plastilab™ Conical Stoppers
Features tapered conical-shaped bottom to ensure sealing action.
Brand: Kartell™ 0124600
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Made from Polyethylene for tight and secure seal
- Raised embossed easy-to-read sizes
- Color-coded
Specifications
Conical bottom | |
Stopper | |
Polyethylene | |
DIN 12242 |
Black | |
25.8 mm | |
NS 29/32 | |
Pentagonal top |