Learn More
Buerkle™ Plastic Single-use Pipets
Product Code. 11832293 Shop All Bürkle Products
Change view
Click to view available options
Volume (Metric):
25 mL
50 mL
100 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11832293 50 mL Each
17210684 25 mL Each
11842293 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11832293 Supplier Buerkle™ Supplier No. 75560050
Only null left
Add to basket
Add to basket
Pipettes made of plastic are particularly suited for training purposes, in industrial labs, during field research and applications where pipettes made of glass cannot be used, e.g. in flourine and silicate chemistry
  • Polystyrene, crystal clear
  • For aqueous solutions
  • |Polystyrene, crystal clear
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
Sterility Sterile
For Use With (Application) For Training Purposes, in Industrial Labs, During Field Research and Applications Where Pipettes Made of Glass Cannot be Used
Quantity Each
Color Crystal Clear
Volume (Metric) 50 mL
Type Disposable Pipette
Disposable Single-use