Learn More
Sartorius Plastic Screw Box for Weights
Product Code. 16333136 Shop All Sartorius Products
Change view
Click to view available options
Holds:
100 g Weights
2 kg Weights
50 g Weights
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
16333136 2 kg Weights Each
15695889 50 g Weights Each
15685889 100 g Weights Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16333136 Supplier Sartorius Supplier No. YAW2000GL
Only null left
Add to basket
Add to basket
Plastic Screw Boxes for preventing any damage to the surface of weights.
  • Plastic screw boxes with foam inlet to be used for weights.
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
For Use With (Application) For use in cleanrooms
Holds 2 kg Weights