missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plastic Open-Top Blue Drum 220L UN Certified
Click to view available options
Capacity (Metric):
220 L
Description
Full open top heavy duty plastic drums are supplied with a plastic lid that is held in place by tamper evident steel closing ring. Manufactured from food grade material. All drums are UN approved to carry hazardous materials.
- Manufactured from food grade material.
- Removable plastic lid with internal gasket and steel tamper evident closing ring.
- Blue standard colour.
- UN approved for transportation of hazardous materials.
Specifications
Specifications
| Capacity (Metric) | 220 L |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction