Learn More
Lenz Laborglasintrumente™ Plastic Octagonal Plate Stoppers
Description
- DIN 12254
- Pack of 10
Specifications
Specifications
| European Size No. | NS 12/21 |
| Quantity | 10 |
| Standard Taper Joint | NS 12/21 |
| Color | White, Red |
| Material | Polyethylene |