missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One EasyLoad™ 200 Refill System

Product Code. 11719482
Change view
Click to view available options
Color:
Natural
Sterility:
Non-sterile
Unit Size:
Pack of 5760
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility unitSize
11719482 Natural Non-sterile Pack of 5760
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11719482 Supplier Greiner Bio-One Supplier No. 741000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Refill system for standard pipette tips, Universal 200μL, thin walled tips with graduation

  • PP (Polypropylene) tips
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Sterility Non-sterile
Color Natural
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line EasyLoad
Material Polypropylene
Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.