missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Plastic Desiccator

Product Code. 10599973 Shop All Bürkle Products
Change view
Click to view available options
Diameter (Metric) Inner:
150 mm
200 mm
250 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10599973 250 mm Each
10182391 200 mm Each
17250654 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10599973 Supplier Buerkle™ Supplier No. 97910250
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Desiccators made of plastic, break resistant, lightweight and suitable for vacuums. Stopcock suitable for vacuums up to 980 mbar. Base and desiccator plate made out of Polypropylene. Lid is crystal clear polycarbonate with a non-return valve and a venting valve. O-ring seal made of polychlorobutadiene-rubber. Continuous temperature max. +60 °C.

  • Hose connection fits hoses with inner Ø 8.5 mm
  • Delivery incl. O-ring
  • Perforated plate and additional O-rings available as accessories
TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene, Polycarbonate
Product Type Desiccator
Includes Polycarbonate lid, Perforated Polypropylene plate and O-ring
Height (Metric) 300 mm
Diameter (Metric) Inner 250 mm
Diameter (Metric) Outer 285 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.