Learn More
Witeg™ Plastic-Coated Round Bottom Flasks
Description
- With spherical ball joint
- Plastic-coated
- Center necks
- Provided in packs of 10
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Glass |
| For Use With (Application) | Boiling |
| Neck Style | Center |
| Socket Joint | S 35/20 |
| Bottom Shape | Round |