missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasmolipin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14868-1-AP-20UL
This item is not returnable.
View return policy
Description
Plasmolipin is an 18 kDa membrane-bound proteolipid that belongs to the tetraspan (4TM) family, a rapidly growing group of myelin-associated proteins. Many of these proteins could be linked to human hereditary demyelinating neuropathies. Studies indicate that plasmolipin is a component of the cytoskeletal structure and has been localized to the apical surface of tubular epithelial cells. It has been shown that, upon addition to lipid bilayers, plasmolipin induces formation of ion channels that are both voltage-dependent and potassium-specific. Plasmolipin was originally isolated from kidney tissue. Subsequently, it has been shown to be widely expressed, not only being present in kidney, but also in the nervous system, thymus, testis, lung, and thyroid gland.Specifications
| Plasmolipin | |
| Polyclonal | |
| Unconjugated | |
| PLLP | |
| Plasma membrane proteolipid, Plasmolipin, PLLP, PMLP, TM4SF11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51090, 64364, 67801 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P47987, Q9DCU2, Q9Y342 | |
| PLLP | |
| Plasmolipin Fusion Protein Ag6425 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction