missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Plasmodium falciparum HRP-2 Monoclonal Antibody (H36F)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518243
This item is not returnable.
View return policy
Description
This antibody recognizes histidine rich protein II (HRP2) of P. falciparum. Purity is >90%. Recommended pairs for ELISA: MA5-18245 or MA5-18246 (capture) and MA5-18243 (conjugated, detection).
Specifications
| Plasmodium falciparum HRP-2 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Protozoa | |
| Antibody | |
| IgG2a |
| ELISA | |
| H36F | |
| Unconjugated | |
| P. falciparum; P. falciparum HRP2; P. falciparum HRP-2 | |
| Plasmodium falciparum recombinant protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction