missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasmodium Aldolase Rabbit anti-Parasite, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against Plasmodium Aldolase.
Brand: Abnova PAB29676.1mg
This item is not returnable.
View return policy
Specifications
| Plasmodium Aldolase | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Plasmodium Aldolase. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| Recombinant protein corresponding to Plasmodium Aldolase. | |
| 1 mg | |
| Primary | |
| Parasite | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (2 ug/mL) Western Blot The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| Antigen Affinity Purification | |
| RUO | |
| Plasmodium aldolase. | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction