missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Plasminogen Monoclonal Antibody (H16), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543960
This item is not returnable.
View return policy
Description
Plasminogen is a plasma protein synthesized mostly in the liver. It consists of a single polypeptide chain and has a molecular weight of 88.5 kDa. The plasma concentration is usually in the range of 70 - 200 mg/L. Plasminogen is activated by tissue-plasminogen activator, urokinase and streptokinase, to form plasmin. Activation results from the cleavage and release of the preactivation peptide. Plasmin consists of a heavy (A-) chain which contains 5 kringles and a light (B-) chain with the active site. Plasmin is a trypsin-like protease whose natural substrate is fibrin, with binding sites for fibrin residing on the kringles. Plasmin is essential for fibrinolysis and decreased fibrinolytic potential due to congenital defects in plasmin can lead to recurring thrombosis, thrombophlebitis and pulmonary embolism.
Specifications
| Plasminogen | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P00747 | |
| PLG | |
| Native protein, with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| H16 | |
| Unconjugated | |
| PLG | |
| Ab1-346; Activation peptide; AI649309; angiostatin; DKFZp779M0222; Pg; plasmin; plasmin heavy chain A; Plasmin heavy chain A, short form; Plasmin light chain B; plasminogen; PLG; RP1-81D8.1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 5340 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction