missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Plasminogen Activator Inhibitor 1 Human Chromogenic Assay Kit, 100 Tests
Shop All Abcam ProductsDescription
- Applications: Enzyme activity assay
- Detection method: Colorimetric
- Range: 0.625 to 20 IU/mL
- Sample types: Cell culture supernatant, Plasma, Serum
- Sensitivity: 0.56 AU/mL
- Results type: Quantitative
- Instrument compatibility: Microplate reader
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Western Blot Kit |
| Detection Location | In-Solution Detection |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 100 Tests |
| Reactivity | Human |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Plasminogen Activator Inhibitor Type 1 (Human) |
| Detection Method | Colorimetric |
| Cross Reactivity | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?