missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- PlasmidSelect Xtra allows for generic purification of supercoiled plasmid DNA
- Allows for consistent purification results when scaling up from research scale to cGMP production
- Convenient formats for research, process development and quantitative and qualitative analysis
- The hydrophilic nature of the base matrix ensures low levels of non-specific binding
- This resin is a BioProcess medium supported for industrial applications
- Composed of 6% highly cross-linked agarose
Specifications
Specifications
| Quantity | 200 mL |
| Stability | Stable in all commonly used aqueous buffers |
| For Use With (Application) | Generic Purification of Supercoiled Plasmid DNA |
| Storage Requirements | 4 to 30 deg.C, 20% Ethanol |
| Product Type | PlasmidSelect Xtra Media |
| Particle Size | 24 to 44 μm |
| Grade | BioProcess resin |
| Max. Pressure | <120 cm/h |
| Matrix | Cross-linked Agarose |
| pH Range | 3 to 11 |
| Show More |
Safety and Handling
Product Identifier
- PlasmidSelect Xtra, 200 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?