missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Buffer A4 Concentrate
Description
The eight-well format of the NucleoSpin™ miniprep columns is easily processed by vacuum filtration. The NucleoVac 96 vacuum manifold allows simultaneous processing of multiples of eight samples in less than 90min. The NucleoSpin™ Multi-8 Plus Plasmid kit also contains NucleoSpin™ Plasmid filter strips to clear bacterial lysates instead of time consuming centrifugation. The NucleoSpin™ Multi-8 Plus Plasmid kit is designed for purifying up to 20μg of high copy-number plasmid DNA from 1mL to 5mLE. coliovernight cultures preferably grown in LB medium. The NucleoSpin™ Plasmid binding strips contain a unique silica membrane that binds DNA in the presence of chaotropic salt. After a modified alkaline lysis procedure, bacterial lysates are loaded onto the NucleoSpin™ Plasmid filter strips arranged on the NucleoVac 96 vacuum manifold. Filtrates are collected directly in the NucleoSpin™ Plasmid binding strips. After binding and washing highly pure plasmid DNA is eluted in low-salt buffer (5mM Tris/HCl, pH 8.5). Alternatively, water (pH 8.5) can be used. Kit components: NucleoSpin™ Plasmid filter strips, NucleoSpin™ Plasmid binding strips, buffers, RNase A, MN wash plate, MN tube strips, cap strips. For further information on this product contact your local Customer Services.
- Silica-membrane technology
- Yield: up to 20μL sequencing grade plasmid DNA
- Elution volume: 75μL to 150μL
- Culture volume: 1mL to 5mL
- Binding capacity: 25μg
- Maximum plasmid size <15kb
- No use of organic solvents
- NucleoSpin 8 filter strips for convenient filtration of bacterial lysates included Rapid small-scale preparation of high-copy plasmids fromE. coli.
Specifications
Specifications
| Chemical Name or Material | Wash Buffer A4 Concentrate |
| For Use With (Equipment) | For 1000mL Buffer A4 |
| Quantity | 200 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?