missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Lysis Buffer LYS
Specifications
Specifications
| Chemical Name or Material | Lysis Buffer |
| For Use With (Application) | For Plasmid DNA Purification |
| Quantity | 1000 mL |
Safety and Handling
Signal Word
- Warning
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contain: sodium hydroxide, dodceyl sulphate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?