missing translation for 'onlineSavingsMsg'
Learn More

Plasmid DNA isolation solution I, Thermo Scientific™

£21.40 - £48.35

Specifications

Chemical Name or Material Plasmid DNA isolation solution I
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15433879
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J61263.AK
250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15443879
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J61263.AP
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Plasmid DNA isolation solution I
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications