missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasma Kallikrein/KLKB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58978
This item is not returnable.
View return policy
Description
Plasma Kallikrein/KLKB1 Polyclonal specifically detects Plasma Kallikrein/KLKB1 in Human samples. It is validated for Western Blot.
Specifications
| Plasma Kallikrein/KLKB1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 3.4.21, EC 3.4.21.34, Fletcher factor, kallikrein B, plasma (Fletcher factor) 1, kininogenin, KLK3plasma kallikrein, plasma kallikrein heavy chain, plasma kallikrein light chain, Plasma prekallikrein, PPK | |
| Rabbit | |
| 28 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Rat: 79%. | |
| Human, Rat, Pig, Canine, Equine | |
| Purified |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P03952 | |
| KLKB1 | |
| Synthetic peptides corresponding to KLKB1(kallikrein B, plasma (Fletcher factor) 1) The peptide sequence was selected from the middle region of KLKB1. Peptide sequence VLTAAHCFDGLPLQDVWRIYSGILNLSDITKDTPFSQIKEIIIHQNYKVS. | |
| Protein A purified | |
| RUO | |
| 3818 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction