missing translation for 'onlineSavingsMsg'
Learn More

Plasma Kallikrein/KLKB1 Antibody (736811) [PE], Novus Biologicals™

Product Code. 30736978 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736978 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736978 Supplier Novus Biologicals Supplier No. FAB2497P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Plasma Kallikrein/KLKB1 Monoclonal antibody specifically detects Plasma Kallikrein/KLKB1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plasma Kallikrein/KLKB1
Applications Immunohistochemistry
Classification Monoclonal
Clone 736811
Conjugate PE
Formulation PBS
Gene Alias EC 3.4.21, EC 3.4.21.34, Fletcher factor, kallikrein B, plasma (Fletcher factor) 1, kininogenin, KLK3plasma kallikrein, plasma kallikrein heavy chain, plasma kallikrein light chain, Plasma prekallikrein, PPK
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human KLK-B1, Gly20-Ala638, Accession # P03952
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3818
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.