missing translation for 'onlineSavingsMsg'
Learn More

Plasma Emission Standards Kit 1 For ICP, SPEX CertiPrep™

Product Code. 12883266 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
38 x 125 mL
Unit Size:
4.8kg
This item is not returnable. View return policy

Product Code. 12883266

Brand: SPEX Certiprep ICPKIT1

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Assurance™ Grade Standards Kit for AA & ICP, 1,000 μg/mL. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Assurance™ Grade Standard Kit for AA & ICP
  • 1,000 μg/mL Concentration
  • 125 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Plasma Emission Standards Kit 1
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Quantity 38 x 125 mL
Concentration or Composition (by Analyte or Components) Each component in a separate 100mL bottle at 1000μg/mL.Component in 20% HCl (in PTFE bottle):TinComponents in 10% HNO3:BismuthMercuryComponents in 2% HNO3:AluminumArsenicBariumBerylliumCadmiumCalciumChromiumCobaltCopperIronLeadLithiumMagnesiumManganeseNickelPotassiumScandiumSeleniumSilverSodiumStrontiumThalliumVanadiumYttriumZincZirconiumComponents in H2O:AntimonyBoronMolybdenumNiobiumPhosphorusSiliconSulfurTitaniumTungsten

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.