missing translation for 'onlineSavingsMsg'
Learn More

Plasma Cell Marker Antibody (SPM310), Novus Biologicals™

Product Code. 18406201 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
Unit Size:
0.02mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406201 0.1 mg 0.1mg
18419951 0.02 mg 0.02mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18406201 Supplier Novus Biologicals Supplier No. NBP2343590.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Plasma Cell Marker Monoclonal antibody specifically detects Plasma Cell Marker in Human, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plasma Cell Marker
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM310
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 0.1-0.2 ug/mL
Formulation 10 mM PBS with 0.05% BSA
Host Species Mouse
Immunogen Pancreatic cancer-related mucin
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rat (Negative)
Content And Storage Store at 4C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.