missing translation for 'onlineSavingsMsg'
Learn More

EduLab™ Plane Glass Mirrors

Product Code. 13172930
Click to view available options
Description:
Dimensions: 100 x 75 mm
Dimensions: 150 x 100 mm
Dimensions: 150 x 25 mm
Dimensions: 150 x 50 mm
Dimensions: 75 x 25 mm
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 13172930

Brand: EduLab™ PH0514A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Silver backed with protective coating.

  • Optically worked glass
  • Unmounted
  • Supplied in packs of 10

Specifications

Description Dimensions: 75 x 25 mm
Quantity 10 Ea.
Product Type Plane Glass Mirror
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.