Learn More
Specac™ Plane Anvil for Silver Gate™ ATR and Golden Gate™
Description
Stainless Steel large anvil for Germanium Top-Plate for use in the Golden Gate Single Reflection ATR Accessory GS10500.
Specifications
Specifications
| For Use With (Equipment) | Golden Gate Single Reflection ATR Accessory GS10500 |
| Type | Plane Anvil |