Learn More
Medline Scientific™ Plan Objective
Product Code. 12613035
Change view
Click to view available options
Magnification Power:
0.5X
2X
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Power unitSize
12613035 0.5X Each
12835101 2X -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12613035 Supplier Medline Scientific™ Supplier No. 7700.0005
For use with Varizoom binocular microscopes.
  • 0.5x increases working distance and increases field of view
  • 2.0 x magnification
Specifications
Type Plan Objective
For Use With (Equipment) Varizoom Series Stereomicroscopes
Magnification Power 0.5X
Quantity 1 Ea.