missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plakophilin 4 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37691DL594
This item is not returnable.
View return policy
Description
Plakophilin 4 Polyclonal antibody specifically detects Plakophilin 4 in Human samples. It is validated for Western Blot
Specifications
| Plakophilin 4 | |
| Polyclonal | |
| Western Blot | |
| catenin 4, FLJ42243, p0071FLJ31261, plakophilin 4, plakophilin-4 | |
| This antibody maps to a region between residue 1 to 50 of human Plakophilin 4 using the numbering given in entry NP_003619.2 (GeneID 8502). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8502 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction