missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plakophilin-3 Antibody (23E3/4), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97674C
This item is not returnable.
View return policy
Description
Plakophilin-3 Monoclonal specifically detects Plakophilin-3 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Plakophilin-3 | |
| Monoclonal | |
| DyLight 650 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PKP3 | |
| Clone 23E3/4 is a mouse monoclonal IgG2b antibody derived by fusion of mouse myeloma cells with spleen cells from a mouse immunized with a synthetic peptide corresponding to amino acid residues 570-589 of human plakophilin-3 coupled to keyhole limpet hemocyanin via an additional cysteine residue at the N-terminus (CFTPQSRRLRELPLAADALTF). | |
| Primary | |
| Human, Mouse, Canine | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 23E3/4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| plakophilin 3, plakophilin-3 | |
| Mouse | |
| 0.1 mL | |
| 11187 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction