missing translation for 'onlineSavingsMsg'
Learn More

PLAGL2 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18430566 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430566 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430566 Supplier Novus Biologicals Supplier No. NBP281930FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLAGL2 Polyclonal antibody specifically detects PLAGL2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLAGL2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias C2H2-type zinc finger protein, FLJ23283, KIAA0198, pleiomorphic adenoma gene-like 2, Pleiomorphic adenoma-like protein 2, zinc finger protein PLAGL2, ZNF900
Host Species Rabbit
Immunogen PLAGL2 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human PLAGL2. The immunogen is located within amino acids 40 - 90 of PLAGL2. Amino Acid Squence: NGEKLRPHSLPQPEQRPYS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 5326
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.