missing translation for 'onlineSavingsMsg'
Learn More

PLAGL1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18384206 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384206 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384206 Supplier Novus Biologicals Supplier No. NBP306208AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLAGL1 Polyclonal specifically detects PLAGL1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLAGL1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp781P1017, LOT-1, LOT1ZACLost on transformation 1, MGC126275, MGC126276, PLAG-like 1, pleiomorphic adenoma gene-like 1, pleiomorphic adenoma gene-like protein 1, Pleiomorphic adenoma-like protein 1, Tumor supressor ZAC, ZAC tumor supressor, ZAC1, zinc finger protein PLAGL1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PLAGL1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5325
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.