missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Placental Lactogen/CSH1 Antibody (OTI2G4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73457APC
This item is not returnable.
View return policy
Description
Placental Lactogen/CSH1 Monoclonal antibody specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Placental Lactogen/CSH1 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI2G4 | |
| Western Blot, Immunohistochemistry | |
| Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, Placental lactogen, PLchoriomammotropin | |
| Full length human recombinant protein of human CSH1(NP_072166) produced in HEK293T cell. | |
| 0.1 mL | |
| Cytokine Research, Signal Transduction | |
| 1442 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction