missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Placental Alkaline Phosphatase Recombinant Rabbit Monoclonal Antibody (24GB5430)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557332
This item is not returnable.
View return policy
Description
Placental Alkaline Phosphatase Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Placental Alkaline Phosphatase plays an important role in the regulation of specific inflammatory disease processes. There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney for this form of alkaline phosphatase have been well characterized. Placental Alkaline Phosphatase reacts with a membrane-bound isoenzyme (Regan and Nagao type) of Placental Alkaline Phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. Placental Alkaline Phosphatase is useful in the identification of testicular germ cell tumors. Unlike germ cell tumors, PLAP-positive somatic cell tumors uniformly express epithelial membrane antigen (EMA). A proposed function of Placental Alkaline Phosphatase is matrix mineralization; however, mice that lack a functional form of this enzyme show normal skeletal development. Placental Alkaline Phosphatase has been linked directly to hypophosphatasia, a disorder that is characterized by hypercalcemia and includes skeletal defects. The character of hypophosphatasia can vary, however, depending on the specific mutation since this determines age of onset and severity of symptoms.
Specifications
| Placental Alkaline Phosphatase | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ALPP | |
| Alkaline phosphatase placental type; Alkaline phosphatase Regan isozyme; alkaline phosphatase, placental; alkaline phosphatase, placental type; alkaline phosphomonoesterase; ALP; Alp1; ALPP; AP-TNAP; DOA1; FLJ11281; FLJ40094; FLJ61142; FLJ93059; Germ-cell alkaline phosphatase; glycerophosphatase; HOPS; MGC161443; MGC167935; nagao Isozyme; P PLAP; PALP; PLA2P; PLAA; placental alkaline phosphatase 1; placental heat-stable alkaline phosphatase; PLAP; PLAP1; PLAP-1; rCG23846-like; Regan isozyme; TNAP; TNSALP | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 250 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5430 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05187 | |
| ALPP | |
| A synthesized peptide derived from human Placental alkaline phosphatase (phospho LAP) (aa 181-198). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction