missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Placental Alkaline Phosphatase Monoclonal Antibody (3E5)

Placental Alkaline Phosphatase Monoclonal Antibody (3E5)
Brand: Invitrogen MA515652
This item is not returnable.
View return policy
Description
Placental Alkaline Phosphatase Monoclonal Antibody for Western Blot, IHC, ELISA
Specifications
| Placental Alkaline Phosphatase | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| P05187 | |
| ALPP | |
| Purified recombinant fragment of human ALPP expressed in E. Coli. | |
| RUO | |
| 250 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3E5 | |
| Unconjugated | |
| ALPP | |
| Alkaline phosphatase placental type; Alkaline phosphatase Regan isozyme; alkaline phosphatase, placental; alkaline phosphatase, placental type; alkaline phosphomonoesterase; ALP; Alp1; ALPP; AP-TNAP; DOA1; FLJ11281; FLJ40094; FLJ61142; FLJ93059; Germ-cell alkaline phosphatase; glycerophosphatase; HOPS; MGC161443; MGC167935; nagao Isozyme; P PLAP; PALP; PLA2P; PLAA; placental alkaline phosphatase 1; placental heat-stable alkaline phosphatase; PLAP; PLAP1; PLAP-1; rCG23846-like; Regan isozyme; TNAP; TNSALP | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering