missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLAC4 Polyclonal Antibody

Description
A suggested positive control is human placenta lysate. PA5-21047 can be used with blocking peptide PEP-1161.
PLAC4 (placenta-specific protein 4), was initially discovered as a gene whose mRNA was expressed in placental syncytiotrophoblast and choriocarcinoma cells. While the function of PLAC4 is currently unclear, its gene is known to be located on chromosome 21q22.3. Due to the PLAC4 gene location on chromosome 21 and the release of its mRNA transcripts into maternal bloodstream from the placenta, it has been proposed to be a useful marker noninvasive prenatal chromosome 21 aneuploidy detection, although other reports suggests that PLAC4 mRNA levels are not altered in the maternal circulation of pregnancies with trisomy 21.
Specifications
Specifications
| Antigen | PLAC4 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | PLAC4 |
| Gene Accession No. | Q8WY50 |
| Gene Alias | C21orf115; PLAC4; placenta specific 4; placenta-specific 4; placenta-specific protein 4; PRED78 |
| Gene Symbols | PLAC4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?