missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PLAC2 Polyclonal antibody specifically detects PLAC2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | PLAC2 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ90734, LINC00036, NCRNA00036, placenta-specific 2 (non-protein coding), PP10933 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 17 amino acid synthetic peptide near the center of human PLAC2. The immunogen is located within amino acids 60 - 110 of PLAC2. Amino Acid Squence: WAARGHRKKRSTEGAGE |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?