missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLAA Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554299
This item is not returnable.
View return policy
Description
Immunogen sequence: TGAGRYVPGS ASMGTTMAGV DPFTGNSAYR SAASKTMNIY FPKKEAVTFD QANPTQILGK LKELNGTAPE EKKLTEDDLI LLEKILSLIC Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
Phospholipase A-2-activating protein (PLAA), also named PLAP, can interact with ubiquitin and is involved in the maintenance of ubiquitin levels. PLAA contains multiple consensus domains including WD repeats, PFU domain and PUL domain. Although the function of PLAA remains unclear, ubiquitin binding of PLAA might be the central role that connects ubiquitination and degradation in ERAD. Recent finding revealed that PLAA served as a novel nociceptive mediator after incision, and its expression level is regulated by miR-203.
Specifications
| PLAA | |
| Polyclonal | |
| Unconjugated | |
| Plaa | |
| 2410007N06; AI536418; AU018445; AW208417; D4Ertd618e; DOA1; DOA1 homolog; phospholipase A2 activating protein; phospholipase A2, activating protein; phospholipase A-2-activating protein; phospholipase A2-activating protein; PLA2P; Plaa; PLAP; Ufd3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116645, 18786, 9373 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P27612, P54319, Q9Y263 | |
| Plaa | |
| Recombinant protein corresponding to Human PLAA. Recombinant protein control fragment (Product #RP-92984). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction