missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLA2R1 Recombinant Rabbit Monoclonal Antibody (7I4Z1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555526
This item is not returnable.
View return policy
Description
Immunogen Sequence: YQYDVPWLFY QDAEYLFHTF ASEWLNFEFV CSWLHSDLLT IHSAHEQEFI HSKIKALSKY GASWWIGLQE ERANDEFRWR DGTPVIYQNW DTGRERTVNN Q.
Receptor for secretory phospholipase A2 (sPLA2). Acts as a receptor for phosholipase sPLA2-IB/PLA2G1B but not sPLA2-IIA/PLA2G2A. Also able to bind to snake PA2-like toxins. Although its precise function remains unclear, binding of sPLA2 to its receptor participates in both positive and negative regulation of sPLA2 functions as well as clearance of sPLA2. Binding of sPLA2-IB/PLA2G1B induces various effects depending on the cell type, such as activation of the mitogen-activated protein kinase (MAPK) cascade to induce cell proliferation, the production of lipid mediators, selective release of arachidonic acid in bone marrow-derived mast cells. In neutrophils, binding of sPLA2-IB/PLA2G1B can activate p38 MAPK to stimulate elastase release and cell adhesion.
Specifications
| PLA2R1 | |
| Recombinant Monoclonal | |
| 0.92 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3 | |
| Q13018, Q62028 | |
| Pla2r1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 7I4Z1 | |
| Unconjugated | |
| Pla2r1 | |
| 180 kDa secretory phospholipase A2 receptor; CLEC13C; C-type lectin domain family 13 member C; mannose receptor, C type 2; Mrc2; M-type receptor; phospholipase A2 receptor 1; phospholipase A2 receptor 1, 180kD; phospholipase A2 receptor 1, 180kDa; phospholipase A2, group IB, pancreas, receptor; Pla2g1br; PLA2G1R; PLA2-I receptor; PLA2IR; Pla2r; Pla2-r; PLA2R1; Secretory phospholipase A2 receptor; Soluble PLA2-R; Soluble secretory phospholipase A2 receptor | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18779, 22925, 295631 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction