missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2R1 Antibody (PLA2R1/4893), Novus Biologicals™
Mouse Monoclonal Antibody
£195.00 - £368.00
Specifications
| Antigen | PLA2R1 |
|---|---|
| Clone | PLA2R1/4893 |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30288541
|
Novus Biologicals
NBP3-44596-100ug |
100 μg |
£368.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30297835
|
Novus Biologicals
NBP3-44596-20ug |
20 μg |
£195.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLA2R1 Monoclonal antibody specifically detects PLA2R1 in Human samples. It is validated for ImmunohistochemistrySpecifications
| PLA2R1 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 22925 | |
| IgG2 κ | |
| Protein A purified |
| PLA2R1/4893 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| 180 kDa secretory phospholipase A2 receptor, CLEC13CC-type lectin domain family 13 member C, phospholipase A2 receptor 1, 180kDa, PLA2G1R, PLA2IR, PLA2-RM-type receptor, PLA2Rphospholipase A2 receptor 1, 180kD, secretory phospholipase A2 receptor | |
| Recombinant fragment (around aa300-600) of human PLA2R1 protein (Uniprot: Q13018). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title