missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2R1 Antibody (12-6-5) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80912
This item is not returnable.
View return policy
Description
PLA2R1 Monoclonal specifically detects PLA2R1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunocytochemistry, CyTOF-ready.
Specifications
| PLA2R1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot : 1:5000 - 1:10000, Flow Cytometry : 1:200, ELISA 1:5000 - 1:20000, Immunohistochemistry : 1:5000 - 1:20000, Immunocytochemistry/Immunofluorescence : 1:10 - 1:500, Immunocytochemistry, CyTOF-ready | |
| 180 kDa secretory phospholipase A2 receptor, CLEC13CC-type lectin domain family 13 member C, phospholipase A2 receptor 1, 180kDa, PLA2G1R, PLA2IR, PLA2-RM-type receptor, PLA2Rphospholipase A2 receptor 1, 180kD, secretory phospholipase A2 receptor | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 22925 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF | |
| 38692 | |
| Unconjugated | |
| PBS | |
| PLA2R1 | |
| Human PLA2R extracellular sequence N-C3 (containing domains N-terminal Cysteine rich-FibII-CTLD1-CTLD2-CTLD3). Genbank acc no. U17033; amino acids 20-663 Recombinant protein expressed in HEK 293-EBNA-1 cells | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction