missing translation for 'onlineSavingsMsg'
Få mere at vide

PLA2G7/PAF-AH/Lp-PLA2 Antibody [DyLight 755], Novus Biologicals™

Artikelnummer. 30734657 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.1 mL
Pakningsstørrelse:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30734657 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30734657 Leverandør Novus Biologicals Leverandørnr. AF5106Z

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Goat Polyclonal Antibody

PLA2G7/PAF-AH/Lp-PLA2 Polyclonal antibody specifically detects PLA2G7/PAF-AH/Lp-PLA2 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen PLA2G7/PAF-AH/Lp-PLA2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias 2-acetyl-1-alkylglycerophosphocholine esterase, EC 3.1.1, EC 3.1.1.47,1-alkyl-2-acetylglycerophosphocholine esterase, Group-VIIA phospholipase A2, gVIIA-PLA2, LDL-associated phospholipase A2, LDL-PLA(2), LDL-PLA2, lipoprotein-associated phospholipase A2, LP-PLA2, PAF acetylhydrolase, PAFAHPAF 2-acylhydrolase, phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma), platelet-activating factor acetylhydrolase
Host Species Goat
Immunogen E. coli-derived recombinant human PLA2G7/PAF-AH/Lp-PLA2, Met33-Asn441, Accession # AAH38452
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 7941
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.