missing translation for 'onlineSavingsMsg'
Learn More

PLA2G7/PAF-AH/Lp-PLA2 Antibody [DyLight 350], Novus Biologicals™

Product Code. 30732828 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732828 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732828 Supplier Novus Biologicals Supplier No. AF5106D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

PLA2G7/PAF-AH/Lp-PLA2 Polyclonal antibody specifically detects PLA2G7/PAF-AH/Lp-PLA2 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G7/PAF-AH/Lp-PLA2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias 2-acetyl-1-alkylglycerophosphocholine esterase, EC 3.1.1, EC 3.1.1.47,1-alkyl-2-acetylglycerophosphocholine esterase, Group-VIIA phospholipase A2, gVIIA-PLA2, LDL-associated phospholipase A2, LDL-PLA(2), LDL-PLA2, lipoprotein-associated phospholipase A2, LP-PLA2, PAF acetylhydrolase, PAFAHPAF 2-acylhydrolase, phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma), platelet-activating factor acetylhydrolase
Host Species Goat
Immunogen E. coli-derived recombinant human PLA2G7/PAF-AH/Lp-PLA2, Met33-Asn441, Accession # AAH38452
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 7941
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.