missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G6 Antibody (OTI10G4), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73456AF594
This item is not returnable.
View return policy
Description
PLA2G6 Monoclonal specifically detects PLA2G6 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PLA2G6 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PLA2G6 | |
| Recombinant protein fragment corresponding to amino acids 490-752 of human PLA2G6 (NP_001004426) produced in E.coli. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI10G4 | |
| Western Blot | |
| calcium-independent phospholipase A2, EC 3.1.1.4, Group VI phospholipase A2, IPLA2, iPLA285 kDa calcium-independent phospholipase A2, IPLA2-VIA, NBIA2A, NBIA2B, PARK14GVI, patatin-like phospholipase domain containing 9, phospholipase A2, group VI (cytosolic, calcium-independent), PNPLA9 | |
| Mouse | |
| 0.1 mL | |
| 8398 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction