missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLA2G4F Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103776
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PLA2G4F.
Calcium-dependent phospholipase A2 that selectively hydrolyzes glycerophospholipids in the sn-2 position. Has higher enzyme activity for phosphatidylethanolamine than phosphatidylcholine.
Specifications
| PLA2G4F | |
| Polyclonal | |
| Unconjugated | |
| PLA2G4F | |
| 3.1.1.4; 4732472I07Rik; cPLA2-zeta; cytosolic phospholipase A2 zeta; Gm647; Phospholipase A2 group IVF; phospholipase A2, group IVF; Pla2g4f; PLA2G4FZ; Pla2zeta | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 255189, 271844 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q50L41, Q68DD2 | |
| PLA2G4F | |
| A synthesized peptide derived from human PLA2G4F(Accession Q68DD2), corresponding to amino acid residues P758-E808. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction